Learn More
(RS)-N-Despropyl Pramipexole, TRC
Product Code. 30357568
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30357568 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30357568 Supplier Toronto Research Chemicals Supplier No. TRCD4171151G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (6RS)-4,5,6,7-Tetrahydro-1,3-benzothiazole-2,6-diamine
CAS 104617-49-4
Molecular Formula C7 H11 N3 S
Quantity 1 g
Synonym 4,5,6,7-Tetrahydro-2,6-benzothiazolediamine, (RS)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole, 2,6-Diamino-4,5,6,7-tetrahydro-1,3-benzothiazole, 2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole, 4,5,6,7-Tetrahydrobenzothiazole-2,6-diamine
InChI Formula InChI=1S/C7H11N3S/c8-4-1-2-5-6(3-4)11-7(9)10-5/h4H,1-3,8H2,(H2,9,10)
SMILES NC1CCc2nc(N)sc2C1
IUPAC Name 4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine
Molecular Weight (g/mol) 169.25
Formula Weight 169.0674