missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ RS 504393

Product Code. 30001425
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001425 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30001425 Supplier R&D Systems™ Supplier No. 2517/10
  • ONLINE EXCLUSIVE PRICE
    339.00€
    320.25€ / 10mg
    Save 18.75€
    5% Off
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Highly selective CCR2 chemokine receptor antagonist

Extremely selective CCR2 chemokine receptor antagonist (IC50 values are 98 nM and > 100 μM for inhibition of human recombinant CCR2b and CCR1 receptors respectively). Inhibits MCP-1 chemotaxis (IC50 = 330 nM) and ischemia-reperfusion injury in kidneys.
TRUSTED_SUSTAINABILITY

Specifications

CAS 300816-15-3
Quantity 10 mg
Product Line Tocris
Molecular Formula C25H27N3O3
Formula Weight 417.5
Purity >99%
Molecular Weight (g/mol) 417.509
Chemical Name or Material RS 504393
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.