Learn More
Macherey-Nagel™ RS 25 Alox N Flash RS cartridge
Click to view available options
:
I.D.: 21.2mm
Specifications
Specifications
| Length (Metric) | 165 mm |
| Type | Flash RS Cartridge |
| For Use With (Equipment) | Use in The Teledyne Isco CombiFlash Systems |