Learn More
Chemsplash Overshoes (Pairs) PB 6B Elasticated Ankle
Description
Chemsplash Elasticated Overshoe
- Chemsplash 55GSM Type PB 6B Material
- 16 in. in Length
- Silicone and Latex Free
- Non-Linting
- Anti-Static Material
- Conforms to ISO 14644-1:2015 Cleanroom 5 - 9
- PFAS Free
Description: The Chemsplash Elasticated Overshoe is made from Chemsplash soft and microporous fabric. It is strong and resistant to chemical splash and dust. It is anti-static to meet EN1149-5:2008, lint-free, and suitable for cleanroom applications. Conforms to ISO 14644-1:2015 Cleanroom 5 - 9. Chemsplash accessories can be used to protect the wearer from their environment or the environment from the wearer. The range has been designed to allow for maximum comfort and wearability.
Specifications
Specifications
| Size | X-Large |
| Material | Microporous Fabric |
| Latex Free | Yes |
| Color | White |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Closure Type | Elastic Ankle |
| Sole Type | Seamless |
| Resistance | Liquid |
| Product Type | Controlled Environments Shoe Covers |
| Show More |