Learn More
Sartorius RPMI 1640 Medium with L-Glutamine
Product Code. 31908352 Shop All Sartorius Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
31908352 100 mL 100mL
31908385 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31908352 Supplier Sartorius Supplier No. 011001B
Developed for long-term culture of blood cells, including normal and abnormal human leukocytes and hybridoma cultures.
  • Modified with Glutamine
Specifications
Form Liquid
Product Type RPMI Cell Culture Media
With Additives Glutamine
Without Additives None
Quantity 100 mL