missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&DSystems™ RPMI 1640, L-glutamine, no phenol red
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Specifications
Specifications
| For Use With (Application) | Cell Culture |
| Quantity | 500 mL |
| Purity or Quality Grade | 0.2 μm filtered |
| Content And Storage | 2 - 8 °C, protected from light |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction