missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Roxadustat
GREENER_CHOICE

Product Code. 18745416
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18745416 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18745416 Supplier Tocris Bioscience™ Supplier No. 7486/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HIF-PH inhibitor; erythropoietic

Roxadustat is a hypoxia-inducible factor prolyl hydroxylase enzyme (HIF-PH) inhibitor (IC50 = 591.4 nM). Roxadustat reduces basal oxygen consumption and increases glycolysis in vitro. It increases hemoglobin levels and stimulates erythropoiesis in vivo. By reducing ACE2 expression, roxadustat reduces the levels of SARS-CoV-2 viral RNA and inhibits entry, replication and secretion of infectious particles in lung epithelial cells. Roxadustat also suppresses hydroxylation and secretion of high molecular weight forms of mannose-binding lectin. The compound suppresses ion currents in pituitary tumor cells (IC50 values for inhibition of peak and late components of delayed rectifier calcium currents are 5.71 and 1.32 μM, respectively). Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[(4-Hydroxy-1-methyl-7-phenoxy-3-isoquinolinyl)carbonyl]glycine
CAS 808118-40-3
Quantity 50 mg
Target Hydrolase Inhibitors
Molecular Formula C19H16N2O5
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.