Learn More
Testo™ Round Indicators Testoterm
46.80€
Product Code | Brand | Temperature Range (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Temperature Range (Metric) | Price | Quantity & Availability | |||||
10116711
|
Testo™
0646 0073 |
88 to 110°C |
46.80€
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
10399861
|
Testo™
0646 0075 |
143 to 166°C |
46.80€
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
10649851
|
Testo™
0646 0076 |
171 to 193°C |
46.80€
Pack of 10 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||