Learn More
Greiner Bio-One Round Bottom Polypropylene Culture Tube with Two-Position Vent Stopper
Product Code. p-7075363
Click to view available options
Capacity (Metric):
14 mL
No. per Pack:
1
No. per Case:
800
Unit Size:
Pack of 800
Product Code. 10778561
Brand: Greiner Bio-One 187261
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
With graduations.
  • Sterile
TRUSTED_SUSTAINABILITY
Specifications
Closure Material LDPE
Diameter (Metric) 18 mm
Autoclavable Autoclavable (Without Vent Stopper)
Certifications/Compliance Free of detectable DNase/RNase, human DNA and pyrogens
Closure Type Dual-position Vent Stopper
Color Translucent
Length (Metric) 95 mm
Material Polypropylene
Includes Writing area
No. per Pack 1
No. per Case 800
Sterility Sterile
Capacity (Metric) 14 mL