Learn More
Cowie Technology™ Round-Bottom Flask, PTFE
Product Code. 10426982 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
10 mL
25 mL
50 mL
100 mL
Socket Joint:
19/26
24/29
29/32
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Socket Joint unitSize
10426982 10 mL 19/26 Each
10221572 25 mL 24/29 Each
10487552 50 mL 29/32 Each
10794903 100 mL 29/32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10426982
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
COWIE TECHNOLOGY™ Round-Bottom Flask, PTFE has a smooth internal finish and is usable at high temperatures.
  • Round Bottom Flasks have a smooth internal finish.
  • PTFE used in the manufacture of Cowie™ PTFE Laboratory Products is pure, virgin material that conforms to FDA and USP Class VI requirements.
  • Inert and usable to 280°C.
  • Tapers match standard ground glass joints.
  • Without sealing rings.
Specifications
Diameter (Metric) Base 32 mm
Capacity (Metric) 10 mL
Socket Joint 19/26
Temperature Range (Metric) Up to 280°C