Learn More
IKA™ Round Bottom Flask Carrier
Shop All IKA Products
Click to view available options
Capacity (Metric):
100 mL
1000 mL
2000 mL
Unit Size:
Each
Description
- Round/hollow cylindrical bottom
- Aluminum anodized
Specifications
Specifications
| Capacity (Metric) | 100 mL |
| Diameter (English) | 5.6 in. |
| Diameter (Metric) | 142 mm |
| Includes | Without handle |
| Shape | Round Bottom |
| Temperature (Metric) | 180°C |
| Type | Flask Carrier |