Learn More
behr Labor-Technik™ Round Bottom Digestion Vessels
Product Code. 10374972
Change view
Click to view available options
Capacity (Metric):
250 mL
750 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10374972 250 mL Each
10389910 750 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10374972 Supplier behr Labor-Technik™ Supplier No. B00217959
Specifications
Product Type Digestion vessel
Capacity (Metric) 250 mL
For Use With (Equipment) InKjel Infrared Digestion Devices