Learn More
DWK Life Sciences Kimble Chase™ Soda Lime Glass Round Bottom Culture Tube with PP Cap
Shop All DWK Life Sciences ProductsDescription
- Clear soda lime glass
- Round bottom
- Polypropylene cap
Specifications
Specifications
| Capacity (Metric) | 30 mL |
| Length (Metric) | 180 mm |
| Material | Soda Lime Glass |
| Packaging | 100Pack |
| Autoclavable | Autoclavable |
| Diameter (Metric) | 18 mm |
| Closure Type | Screw Cap |
| Product Type | Culture Tube |
| Bottom Shape | Round |