Learn More
Gilac™ Round Bedpan with Lid
Product Code. 17988931
Change view
Click to view available options
Volume (Metric):
2.5 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
17988931 2.5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17988931 Supplier Gilac™ Supplier No. G101421
  • ONLINE EXCLUSIVE PRICE
    20.92€
    13.82€ / Pack of 1
    Save 7.10€
    33% Off
Estimated Shipment: 09-12-2026
Round Bedpan with Lid is suitable for hospital use.
  • Suitable for hospital use.
  • Assembled bedpan and lid, round shape.
  • Outstanding durability even in the intensive use cycle.
  • Product is disinfected in a bedpan washer.
  • Researched product ergonomics ensure optimum cleaning.
  • Equipment at room temperature to avoid the cold sensation experienced with stainless steel equivalents.
Specifications
Product Type Bedpan
Color White
Material Polypropylene
Sterility Non-sterile
Volume (English) 0.7 gal.
Volume (Metric) 2.5 L