Learn More
Thermo Scientific™ mySPIN™ Mini Centrifuge Series Rotors
Shop All Thermo Scientific Products
Click to view available options
Capacity:
12 x 1.5/2 mL
16 x 0.2 mL
32 x 0.2 mL
6 x 1.5/2 mL
Max. RCF:
2,000 x g
9,800 x g
For Use With (Equipment):
mySPIN 12 Mini Centrifuge
mySPIN 6 Mini Centrifuge
Unit Size:
1 unit
Description
Spin 0.2mL and 1.5/2.0mL tubes with Thermo Scientific™ mySPIN™ Mini Centrifuge Series Rotors.
Specifications
Specifications
| Description | 6 x 1.5/2.0mL Circular Rotor |
| Material | Reinforced Glass Fiber |
| Type | Fixed Angle Rotor |
| Capacity | 6 x 1.5/2 mL |
| Max. RCF | 2,000 x g |
| Max. Speed | 6,000 rpm |
| For Use With (Equipment) | mySPIN 6 Mini Centrifuge |
| Biocontainment | No |