Learn More
Corning™ Rotor for LSE™ Mini Microcentrifuge
Corning™ Rotor for Corning™ Mini Microcentrifuge 6770
28.45€ - 40.45€
Specifications
Type | Rotor |
---|---|
For Use With (Equipment) | Corning™ Mini Microcentrifuge 6770 |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
15820982
|
Corning™
6770-RTS |
4 x 8 Strip Tubes |
28.45€
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
15810982
|
Corning™
6770-RT |
8 x 1.5/2.0 mL |
40.45€
Each |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
Description
Corning™ Rotor for Corning™ Mini Microcentrifuge 6770Specifications
Rotor | |
Corning™ Mini Microcentrifuge 6770 |