Learn More
IKA™ ROTAVISC Standard Spindle
Product Code. 17267549 Shop All IKA Products
Click to view available options
Size:
14 x 117 x 14 mm (W x H x D)
16 x 135 x 16 mm (W x H x D)
20 x 117 x 20 mm (W x H x D)
22 x 135 x 22 mm (W x H x D)
32 x 135 x 32 mm (W x H x D)
35 x 135 x 35 mm (W x H x D)
55 x 135 x 55 mm (W x H x D)
8 x 117 x 8 mm (W x H x D)
8 x 135 x 8 mm (W x H x D)
Form:
Cylinder
Disc
Unit Size:
Each
Product Code. 17267549
Brand: IKA™ 0025006839
Standard spindle designed for use with IKA™ ROTAVISC Viscometers.
  • Quick coupling
  • Hook hitch
  • M2.5 left-handed thread
TRUSTED_SUSTAINABILITY
Specifications
Product Type Standard Spindle
For Use With (Equipment) IKA™ ROTAVISC Viscometer
Material AISI 316L
Form Cylinder
Size 8 x 135 x 8 mm (W x H x D)