Learn More
Testo™ Rotating Vane Anemometer
Product Code. 10427173
Click to view available options
Dimensions (L x W x H):
277 x 105 x 45 mm
Unit Size:
Each
Product Code. 10427173
Brand: Testo™ 0560 4170
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Type Vane Anemometer
Accuracy ±0.5 m/sec.
Dimensions (L x W x H) 277 x 105 x 45 mm
For Use With (Application) Measurement of volumetric flow, flow velocity and temperature
Temperature (Metric) -20 to +70°C