Learn More
Olympus™ Rotatable Analyzer, U-AN360-3
Product Code. 12611521
Click to view available options
For Use With (Equipment):
U-RLA, U-URA vertical illuminators
Unit Size:
Each
Product Code. 12611521
Brand: Olympus™ K10017704
Complete polarized visualization with this slider analyzer. Olympus™ Rotatable Analyzer, U-AN360-3 rotates 360° and includes a locking mechanism.
  • Rotatable: 360°.
  • Slide plate.
  • For U-RLA, U-URA vertical illuminators.
Specifications
Type Rotatable
For Use With (Equipment) U-RLA, U-URA vertical illuminators