Learn More
Hirschmann™ Rotarus™ adapter-set PKF 60 for Pump Drives Volume
Click to view available options
Dimensions (L x W x H):
108 x 10 x 89 mm
Unit Size:
Each
Description
- Adapter plate: PFA
- Coupling piece: aluminum
- Connecting PKF 60-series single-channel pump heads with the rotarus™ pump drives volume
Specifications
Specifications
| Color | Black |
| Dimensions (L x W x H) | 108 x 10 x 89 mm |
| Material | POM |
| Overall Height (Metric) | 108 mm |
| Overall Length (Metric) | 10 mm |
| Overall Width (Metric) | 89 mm |
| Quantity | 1 |
| Sterility | Non-sterile |