Learn More
Rotaflo™ Stopcock Spare Parts
Product Code. 12947306
Change view
Click to view available options
Type:
Locking Ring
Stopcock Key
Stopcock Spare Parts
Plug Bore:
3 mm
6 mm
10 mm
Unit Size:
Pack of 5
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Plug Bore Unit Size
12947306 Locking Ring 3 mm Pack of 5
12852785 Locking Ring 6 mm Pack of 5
10213410 Stopcock Key 3 mm Pack of 5
12967306 Stopcock Key 6 mm Pack of 5
12693022 Stopcock Key 10 mm Pack of 5
12643032 Stopcock Spare Parts 10 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12947306 Supplier Quickfit Supplier No. LR3
  • ONLINE EXCLUSIVE PRICE
    8.47€
    6.68€ / Pack of 5
    Save 1.79€
    21% Off
Estimated Shipment: 18-01-2027
Replacement locking rings and keys for Quickfit Rotaflo™ stopcocks
Replacement keys and locking rings should be ordered separately (please see our website, or contact Customer Service)
Specifications
Type Locking Ring
Plug Bore 3 mm
Material Borosilicate Glass
For Use With (Equipment) Rotaflo™ stopcock