missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies RosetteSep™ Human NK Cell Enrichment Cocktail

Product Code. 17137661
Encompass
Change view
Click to view available options
Quantity:
200 mL whole blood
40 mL whole blood
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17137661 40 mL whole blood 1 set
17107671 200 mL whole blood 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17137661 Supplier STEMCELL Technologies Supplier No. 15025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunodensity isolation of untouched NK cells

  • The RosetteSep™ Human NK Cell Enrichment Cocktail is designed to isolate NK cells from whole blood by negative selection
  • Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing non-NK cells and red blood cells (RBCs)
  • When centrifuged over a buoyant density medium such as RosetteSep™ DM-L (Catalog #15705) or Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs
  • The purified NK cells are present as a highly enriched population at the interface between the plasma and the buoyant density medium

Specifications

Cell Type NK Cells
For Use With (Application) Cell Isolation
Quantity 40 mL whole blood
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.