missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies RosetteSep™ CTC Enrichment Cocktail Containing Anti-CD36
Click to view available options
Quantity:
200 mL whole blood
40 mL whole blood
Unit Size:
1 set
Description
- The RosetteSep™ CTC Enrichment Cocktail Containing Anti-CD36 is designed to enrich circulating epithelial tumor cells from fresh whole blood by negative selection
- Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognizing CD2, CD16, CD19, CD36, CD38, CD45, CD66b and glycophorin A on red blood cells (RBCs)
- When centrifuged over a density gradient medium such as Lymphoprep™ (Catalog #07801), the unwanted cells pellet along with the RBCs
- The purified tumor cells are present as a highly enriched population at the interface between the plasma and the density gradient medium
- This cocktail is compatible with small-cell carcinoma samples, including lung cancer samples
- If you are working with breast cancer samples, use the RosetteSep™ CTC Enrichment Cocktail Containing Anti-CD56 (Catalog #15137)
Specifications
Specifications
| Cell Type | Cancer Cells and Cell Lines |
| For Use With (Application) | Cell Isolation |
| Quantity | 200 mL whole blood |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction