Learn More
Thermo Scientific™ Rollout Drawers
Product Code. 13203219 Shop All Thermo Scientific Products
Click to view available options
Type:
Additional SS Drawer
Complete SS Drawer Set
Capacity (English):
11.5 cu. ft.
23.3 cu. ft.
29.2 cu. ft.
78.8 cu. ft.
No. of Drawers:
1
6
7
missing translation for 'unitSize'
1 unit
Each
Pack of 6
Product Code. 13203219
missing translation for 'mfr': Thermo Scientific™ 7446
Only null left
Add to basket
Add to basket
Choose stainless-steel Thermo Scientific™ Rollout Drawers as a convenient alternative to open-wire refrigerator shelving.
Choose stainless-steel Thermo Scientific™ Rollout Drawers as a convenient alternative to open-wire shelving. This is an option for Thermo Scientific™ high-performance refrigerators.
  • Stainless-steel drawers
  • Adjustable with full extension
Ordering Information:
  • Factory installed; must be specified when ordering the refrigerator
TRUSTED_SUSTAINABILITY
Specifications
Type Complete SS Drawer Set
Capacity (English) 11.5 cu. ft.
Capacity (Metric) 326 L
Drawers Stainless Steel (Drawer)
No. of Drawers 7
For Use With (Equipment) 11.5 cu. ft. (326L) Freezers