Learn More
Brady™ Roll Markers
Click to view available options
Display Text:
Air
Eau
Eau incendie
Vapeur
Dimensions (L x W x H):
33 m x 100 mm x 150 mm
33 m x 100 mm x 60 mm
Height (Metric) Character:
8 mm
13 mm
Unit Size:
1 roll
Specifications
Specifications
| Certifications/Compliance | European Directives 67/548 and 88/379 |
| Adhesive Type | Permanent Acrylic |
| Color | White on Green |
| Format | Roll |
| Material | Polyester |
| Product Type | Pipe Marker |
| Display Text | Eau |
| For Use With (Application) | Pipe and valve marking |
| Languages | French |
| No. per Pack | 505 Markers/Roll |
| Show More |