Learn More
Brady™ Roll Markers
Product Code. 11785573 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W): 33m x 100mm; Display Text: Acid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11785573 Dimensions (L x W): 33m x 100mm; Display Text: Acid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11785573 Supplier Brady™ Supplier No. 226773
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Certifications/Compliance European Directives 67/548 and 88/379
Color Purple
Material Polyester
Display Text Acid
For Use With (Application) Used for Pipemarkers and safety signs
Quantity 505/Pk.
Length (Metric) 33 m
Height (Metric) 100 mm
Height (Metric) Character 8 mm