Learn More
Brady™ Roll Marker without Support - Steam
Product Code. 11715563 Shop All Brady Products
Change view
Click to view available options
Width (Metric):
100 mm
Unit Size:
1 roll
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Width (Metric) Unit Size
11715563 100 mm 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11715563 Supplier Brady™ Supplier No. 226759
  • ONLINE EXCLUSIVE PRICE
    340.00€
    306.67€ / 1 rolls
    Save 33.33€
    9% Off
Estimated Shipment: 24-11-2026
Roll Marker without Support (Steam) is made of polyester with polyester overlaminate.
  • Permanent acrylic based adhesive.
Specifications
Adhesive Type Permanent Acrylic
Color Black on Gray
Format Roll
Material Polyester
Product Type Pipe Marker
Display Text Vapeur
For Use With (Application) Pipe and valve marking
Languages French
No. per Pack 220 Markers/Roll
Quantity 220 Pk.
Temperature Range (English) -40°F to +284°F
Temperature Range (Metric) -40°C to +140°C
Thickness (English) 0.0047 in.
Thickness (Metric) 0.119 mm
Dimensions (L x W) 33 m x 100 mm
Dimensions (L x W x H) 33 m x 100 mm x 150 mm
Length (English) 108.27 ft.
Length (Metric) 33 m
Width (English) 3.94 in.
Width (Metric) 100 mm
Height (English) 5.91 in.
Height (Metric) 150 mm
Height (Metric) Character 13 mm
Alternate Part Number PMFL-VAPEUR-GY4-RLL090