Learn More
Rogo Sampaic™ Glass Coverslips
Click to view available options
Length (Metric):
18 mm
20 mm
22 mm
24 mm
Width (Metric):
18 mm
20 mm
22 mm
50 mm
60 mm
Quantity:
100/Pk.
1000/Pk.
200/Pk.
Unit Size:
Pack of 100
Pack of 1000
Pack of 200
Description
- Available in a range of sizes
- Corrosion-free and resistant to thermal shock
Specifications
Specifications
| Product Type | Cover Slide |
| Shape | Square |
| Material | Borosilicate Glass |
| Length (Metric) | 22 mm |
| Width (Metric) | 22 mm |
| Dimensions (L x W) | 22 x 22 mm |
| For Use With (Application) | Slide microscopy |
| Certifications | ISO 8255 |
| Quantity | 200/Pk. |