Learn More
Roflumilast N-Oxide, TRC
Specifications
Specifications
| Chemical Name or Material | Roflumilast N-Oxide |
| CAS | 292135-78-5 |
| Molecular Formula | C17 H14 Cl2 F2 N2 O4 |
| Quantity | 2.5 mg |
| Synonym | 3-(Cyclopropylmethoxy)-N-(3,5-dichloro-1-oxido-4-pyridinyl)-4-(difluoromethoxy)-benzamide','3-(Cyclopropylmethoxy)-N-(3,5-dichloro-1-oxido-4-pyridinyl)-4-(difluoromethoxy)benzamide, N-(3,5-Dichloro-1-oxopyridin-4-yl)-4-difluoromethoxy-3-cyclopropylmethoxybenzamide, Roflumilast N-oxide |
| InChI Formula | InChI=1S/C17H14Cl2F2N2O4/c18-11-6-23(25)7-12(19)15(11)22-16(24)10-3-4-13(27-17(20)21)14(5-10)26-8-9-1-2-9/h3-7,9,17H,1-2,8H2,(H,22,24) |
| SMILES | [O-][n+]1cc(Cl)c(NC(=O)c2ccc(OC(F)F)c(OCC3CC3)c2)c(Cl)c1 |
| IUPAC Name | 3-(cyclopropylmethoxy)-N-(3,5-dichloro-1-oxidopyridin-1-ium-4-yl)-4-(difluoromethoxy)benzamide |
| Molecular Weight (g/mol) | 419.21 |
| Formula Weight | 418.0299 |