Learn More
Cole-Parmer™ Rocking Platform
Product Code. 12014356
Click to view available options
Speed:
15 to 60 rpm
20 to 60 rpm
Unit Size:
Each
Product Code. 12014356
Brand: Cole-Parmer™ 3945516
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Complement your hybridization incubation systems with this Rocking Platform.
  • Allows a number of hybridization tubes to be placed above.
  • Manufactured from stainless steel.
  • Enables membranes to be processed using a wave motion.
  • Speed range of 0 to 60 oscillations per minute.
Specifications
For Use With (Equipment) HI-200 Series Hybridization Incubators
Holds Hybridization Tubes
Material Stainless Steel
Type Rocking Platform
Speed 15 to 60 rpm