missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROCK1 Rabbit anti-Human, Mouse, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
129.00€ - 430.00€
Specifications
Antigen | ROCK1 |
---|---|
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | CoraLite 594 |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17761399
|
Proteintech
CL594-21850-20UL |
20 μL |
129.00€
20µL |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
17679946
|
Proteintech
CL594-21850-100UL |
100 μL |
430.00€
100µL |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
Description
ROCK1 Polyclonal antibody apecifically detetects ROCK1 antigen in Human, Mouse Samples. It is validated for Immunofluorescence.Specifications
ROCK1 | |
Polyclonal | |
Liquid | |
Human, Mouse | |
BC113114 | |
ROCK1 | |
IgG | |
Antigen affinity purification |
Immunofluorescence | |
CoraLite 594 | |
Rabbit | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
6093 | |
ROCK1 fusion protein Ag16317 | |
Primary | |
Store at -20°C. Avoid exposure to light. |