Learn More
Bel-Art™ Robotic Sterile Pipette Tips
Product Code. 17224984 Shop All SP Scienceware Products
Click to view available options
For Use With (Application):
For: Hamilton, Volume: 100 μL, Conductive, Material: carbon filled polypropylene
For: Hamilton, Volume: 100 μL, Non-conductive, Material: Clear polypropylene
For: Hamilton, Volume: 50 μL, Conductive, Material: carbon filled polypropylene
For: Hamilton, Volume: 50 μL, Non-conductive, Material: clear polypropylene
For: Tecan, Volume: 100 μL, Conductive, Material: carbon filled polypropylene
For: Tecan, Volume: 100 μL, Non-conductive, Material: clear polypropylene
For: Tecan, Volume: 50 μL, Conductive, Material: carbon filled polypropylene
For: Tecan, Volume: 50 μL, Non-conductive, Material: clear polypropylene
Unit Size:
Pack of 16
Pack of 24
Product Code. 17224984
Brand: Bel-Art™ F378603005
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Designed for use with automated liquid handling equipment
  • Sterile, disposable, filtered pipette tips can be used in robotic systems
    • Filtered to prevent contamination of pipette arm
  • Conductive tip models detect liquid level when pipetting conductive liquids
    • Ensures repeatable volume accuracy
  • DNase, RNase, pyrogen free, manufactured in Class 8 clean room
  • CE marked
Specifications
Volume (Metric) 50 μL
Filtered Filtered
Sterility Sterile
Color Gray
Autoclavable Not Autoclavable
Certifications/Compliance CE marked
For Use With (Application) For: Hamilton, Volume: 50 μL, Conductive, Material: carbon filled polypropylene
Format Hanging Tip Rack
Tip Style Standard
Packaging 24 racks x 96 tips
Quantity 2304
Product Type Robotic pipette tips
Conductivity Yes
Disposable Single-use
For Use With (Equipment) Hamilton STAR, STARlet and NIMBUS
Graduated No
Material Carbon-filled Polypropylene