Learn More
STEMCELL Technologies RoboSep™ Release Human CD4 Positive Selection Kit
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
1 set
Description
- The EasySep™ Release Human CD4 Positive Selection Kit is designed to isolate CD4+ cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic positive selection
- Desired cells are labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet
- Unwanted cells are simply poured off, while desired cells remain in the tube
- Then, bound magnetic particles are removed from the EasySep™-isolated CD4+ cells, which are immediately available for downstream applications
- Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands
- The CD4 antigen is strongly expressed on helper T cells and is expressed more weakly on peripheral blood monocytes and tissue macrophages
Specifications
Specifications
| Cell Type | T Cells, T Cells, CD4+ |
| Quantity | 1 x 109 cells |