Learn More
STEMCELL Technologies RoboSep™ Release Human CD4 Positive Selection Kit
Product Code. 17187751
Encompass
Click to view available options
Quantity:
1 x 109 cells
Unit Size:
1 set
Product Code. 17187751
Brand: STEMCELL Technologies 17752RF
EasySep™ kit with RoboSep™ buffer and RoboSep™ filter tips
  • The EasySep™ Release Human CD4 Positive Selection Kit is designed to isolate CD4+ cells from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic positive selection
  • Desired cells are labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet
  • Unwanted cells are simply poured off, while desired cells remain in the tube
  • Then, bound magnetic particles are removed from the EasySep™-isolated CD4+ cells, which are immediately available for downstream applications
  • Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands
  • The CD4 antigen is strongly expressed on helper T cells and is expressed more weakly on peripheral blood monocytes and tissue macrophages
Specifications
Cell Type T Cells, T Cells, CD4+
Quantity 1 x 109 cells
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.