missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies RoboSep™ Mouse Streptavidin RapidSpheres™ Isolation Kit
Description
- The EasySep™ Mouse Streptavidin RapidSpheres™ Isolation Kit is designed for the depletion of single or multiple unwanted cell types labeled with biotinylated antibodies
- Labeled cells from mouse splenocytes or other tissues are targeted for removal by Streptavidin RapidSpheres™ and separated without the use of columns using an EasySep™ magnet
- Desired cells are untouched and poured off into a new tube
Specifications
Specifications
| Cell Type | B Cells, Dendritic Cells, Granulocytes and Subsets, Hematopoietic Stem and Progenitor Cells, Macrophages, Marrow Stromal Cells, Mesenchymal Stem and Progenitor Cells, Monocytes, Mononuclear Cells, Myeloid Cells, NK Cells, Other, Plasma, T Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 cells |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?