missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies RoboSep™ Human Monocyte Isolation Kit
Click to view available options
Quantity:
1 x 109 Cells
Unit Size:
1 set
Description
- The EasySep™ Human Monocyte Isolation Kit is designed to isolate CD14+CD16- monocytes from fresh or previously frozen peripheral blood mononuclear cells or washed leukapheresis samples by immunomagnetic negative selection
- The EasySep™ procedure involves labeling unwanted cells and platelets with antibody complexes and magnetic particles
- The magnetically labelled cells are separated from the untouched desired cells by using an EasySep™ magnet and simply pouring or pipetting the desired cells into a new tube
- This product can be used in place of the EasySep™ Human Monocyte Enrichment Kit (Catalog #19059) for even faster cell isolations with reduced platelet contamination
Specifications
Specifications
| Cell Type | Monocytes |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 Cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction