missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies RoboSep™ Human CD14 Positive Selection Kit II
Description
- The EasySep™ Human CD14 Positive Selection Kit II is designed to isolate highly purified CD14+ cells from fresh or previously frozen human peripheral blood mononuclear cells (PBMCs)
- Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD14 and dextran-coated magnetic particles
- Labeled cells are separated using an EasySep™ magnet without the use of columns
- Cells of interest remain in the tube while unwanted cells are poured off
- The CD14 antigen is strongly expressed on monocytes and macrophages and weakly on granulocytes
- It is also expressed on most tissue macrophages
- This product replaces the EasySep™ Human CD14 Positive Selection Kit (Catalog #18058), for even faster cell isolations and is compatible with all EasySep™ magnets
Specifications
Specifications
| Cell Type | Monocytes, Myeloid Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 1 x 109 cells |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?