missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies RoboSep™ Complete Kit for Human Whole Blood CD34+ Cells
Click to view available options
Quantity:
120 mL whole blood
Unit Size:
1 set
Description
- The Complete Kit for Human Whole Blood CD34+ Cells is a simple two-step method that is designed to isolate CD34+ cells from whole blood or buffy coat
- First, hematopoietic progenitor cells are pre-enriched using the RosetteSep™ Human Hematopoietic Progenitor Cell Enrichment Cocktail (15186C) with antibodies recognizing CD2, CD3, CD14, CD16, CD19, CD24, CD56, CD66b and CD61 surface markers
- CD34+ cells are then selected using the EasySep™ Human CD34 Positive Selection Cocktail (18066C), which contains an antibody recognizing CD34
- RosetteSep™ binds unwanted cells to red blood cells (RBCs), forming immunorosettes, which sediment during density gradient centrifugation
- The pre-enriched fraction containing CD34+ cells is harvested and then labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet
- Isolated CD34+ cells are immediately available for downstream applications
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Isolation |
| Quantity | 120 mL whole blood |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction