missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ROBO1 Knockout A549 Cell Lysate

Product Code. 18630064 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18630064 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18630064 Supplier Novus Biologicals™ Supplier No. NBP318631
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Cell Lysate

TRUSTED_SUSTAINABILITY

Specifications

Gene Accession No. Q9Y6N7, Q9Y6N7, Q9Y6N7, Q9Y6N7
Preparation Method This product has been tested by SDS-PAGE and western blot and is suitable for use in Western blot, ELISA, Immunoprecipitation and ChIP. No detection of expected band at ~181 kDa is observed in ROBO1 knockout A549 when compared with unmodified A549 cell lysates by Western blot.
Quantity 100 μg
Purity Multi-step
Host Species Human
Content And Storage Store at -70°C. Avoid freeze/thaw cycles.
Research Category Angiogenesis, Cancer, Extracellular Matrix
Target ROBO1
Synonym DUTT1Drosophila) homolog 1, H-Robo-1, MGC131599, MGC133277, roundabout, axon guidance receptor, homolog 1 (Drosophila)
Gene Symbol ROBO1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.