missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ro 90-7501, TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
Specifications
Specifications
| Chemical Name or Material | 2-[2-(4-aminophenyl)-3H-benzimidazol-5-yl]-3H-benzimidazol-5-amine |
| CAS | 293762-45-5 |
| Molecular Formula | C20 H16 N6 |
| Quantity | 2.5 mg |
| Synonym | Ro 90-7501, 2'-(4-Aminophenyl)-[2,5'-Bi-1H-benzimidazol]-5-amine |
| InChI Formula | InChI=1S/C20H16N6/c21-13-4-1-11(2-5-13)19-23-15-7-3-12(9-17(15)25-19)20-24-16-8-6-14(22)10-18(16)26-20/h1-10H,21-22H2,(H,23,25)(H,24,26) |
| SMILES | Nc1ccc(cc1)c2nc3ccc(cc3[nH]2)c4nc5ccc(N)cc5[nH]4 |
| IUPAC Name | 2-[2-(4-aminophenyl)-3H-benzimidazol-5-yl]-3H-benzimidazol-5-amine |
| Molecular Weight (g/mol) | 340.381 |
| Formula Weight | 340.144 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction