Learn More
Ro 67-4853, TRC
Product Code. 30421356
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421356 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30421356 Supplier Toronto Research Chemicals Supplier No. TRCR63740025MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-(9H-Xanthen-9-ylcarbonyl)carbamic Acid Butyl Ester
CAS 302841-89-0
Molecular Formula C19H19NO4
Quantity 25 mg
Synonym (9H-Xanthen-9-ylcarbonyl)carbamic Acid Butyl Ester, Ro 67-4853
InChI Formula InChI=1S/C19H19NO4/c1-2-3-12-23-19(22)20-18(21)17-13-8-4-6-10-15(13)24-16-11-7-5-9-14(16)17/h4-11,17H,2-3,12H2,1H3,(H,20,21,22)
SMILES CCCCOC(=O)NC(=O)C1c2ccccc2Oc3ccccc13
IUPAC Name butyl N-(9H-xanthene-9-carbonyl)carbamate
Molecular Weight (g/mol) 325.36
Formula Weight 325.1314