Learn More
Ro 67-4853, TRC
Specifications
Specifications
| Chemical Name or Material | N-(9H-Xanthen-9-ylcarbonyl)carbamic Acid Butyl Ester |
| CAS | 302841-89-0 |
| Molecular Formula | C19H19NO4 |
| Quantity | 10 mg |
| Synonym | (9H-Xanthen-9-ylcarbonyl)carbamic Acid Butyl Ester, Ro 67-4853 |
| InChI Formula | InChI=1S/C19H19NO4/c1-2-3-12-23-19(22)20-18(21)17-13-8-4-6-10-15(13)24-16-11-7-5-9-14(16)17/h4-11,17H,2-3,12H2,1H3,(H,20,21,22) |
| SMILES | CCCCOC(=O)NC(=O)C1c2ccccc2Oc3ccccc13 |
| IUPAC Name | butyl N-(9H-xanthene-9-carbonyl)carbamate |
| Molecular Weight (g/mol) | 325.36 |
| Formula Weight | 325.1314 |