missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ RLN1 Recombinant Protein

Brand:  Abnova™ H00006013-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16120232

  • 508.00€ / 25µg
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH05956.1
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
43.12
8
Glutathione Sepharose 4 Fast Flow
25 ug
WKDDVIKLCGRELVRAQIAICGMSTWSKRSLSQEDAPQTPRPVAEIVPSFINKDTETIIIMLEFIANLPPELKAALSERQPSLPELQQYVPALKDSNLSFEEFKKLIRNRQSEAADSNPSELKYLGLDTHSQKKRRPYVALFEKCCLIGCTKRSLAKYC
H1/RLXH1/bA12D24.3.1/bA12D24.3.2
RLN1
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
6013
RLN1 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
RLN1
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers