missing translation for 'onlineSavingsMsg'
Learn More

Rivaroxaban Diol, TRC

Product Code. 30132587
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132587 10 mg 10mg
30137631 25 mg 25mg
17991065 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132587 Supplier Toronto Research Chemicals Supplier No. TRCR53801010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1160170-00-2
Molecular Formula C19H20ClN3O6S
Molecular Weight (g/mol) 453.9
Synonym 2-Thiophenecarboxamide, 5-chloro-N-[[(5S)-3-[4-[(2-hydroxyacetyl)(2-hydroxyethyl)amino]phenyl]-2-oxo-5-oxazolidinyl]methyl]- (ACI), 5-Chloro-N-[[(5S)-3-[4-[(2-hydroxyacetyl)(2-hydroxyethyl)amino]phenyl]-2-oxo-5-oxazolidinyl]methyl]-2-thiophenecarboxamide (ACI), Rivaroxaban Diol, Rivaroxabandiol, Rivaroxaban Open Oxomorpholine Ring, (S)-5-Chloro-N-((3-(4-(2-hydroxy-N-(2-hydroxyethyl)acetamido) phenyl)-2-oxooxazolidin-5-yl)methyl)thiophene-2-carboxamide
IUPAC Name 5-chloro-N-[[(5S)-3-[4-[(2-hydroxyacetyl)-(2-hydroxyethyl)amino]phenyl]-2-oxo-1,3-oxazolidin-5-yl]methyl]thiophene-2-carboxamide
SMILES OCCN(C(=O)CO)c1ccc(cc1)N2C[C@H](CNC(=O)c3ccc(Cl)s3)OC2=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H20ClN3O6S/c20-16-6-5-15(30-16)18(27)21-9-14-10-23(19(28)29-14)13-3-1-12(2-4-13)22(7-8-24)17(26)11-25/h1-6,14,24-25H,7-11H2,(H,21,27)/t14-/m0/s1
Formula Weight 453.0761
Chemical Name or Material Rivaroxaban Diol (Mixture of Diastereomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.