missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ RIPK3/RIP3 Antibody Blocking Peptide
Specifications
Specifications
| Gene ID (Entrez) | 11035 |
| Product Line | Novus |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| For Use With (Application) | This peptide is useful as a blocking peptide for NBP2-24588. For further blocking peptide related protocol, click here . |
| Gene Alias | EC 2.7.11.1, Receptor Interacting Serine/Threonine Kinase 3, Receptor-interacting protein 3, receptor-interacting serine/threonine-protein kinase 3, receptor-interacting serine-threonine kinase 3, RIP3, RIP-3, RIP-like protein kinase 3 |
| Gene Symbol | RIPK3 |
| Quantity | 0.1 mg |
| Research Category | Cancer, Cell Biology, Immunology, Necroptosis, Protein Kinase, Signal Transduction |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?