Learn More
Tocris Bioscience™ RIPK2 inhibitor 1
Description
RIPK2 inhibitor 1 is a potent receptor-interacting protein kinase 2 (RIPK2) inhibitor (IC50 of 5-10 nM). It inhibits NF-κB activity and decreases cancer cell proliferation by 70%. In vivo, RIPK2 inhibitor 1 inhibits intestinal and lung inflammation with low toxicity in rodent models.
Specifications
Specifications
| Chemical Name or Material | 3-(Benzoylamino)-4-methyl-N-[3-(1-methyl-1H-imidazol-2-yl)phenyl]benzamide |
| CAS | 1290490-78-6 |
| Quantity | 10 mg |
| Target | RIP Kinase Inhibitors |
| Molecular Formula | C25H22N4O2 |