Learn More
Elma™ Rinsing Sinks
Product Code. 17861357
Change view
Click to view available options
Model:
L
M
S
XL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
17861357 M Each
17831307 S Each
17821307 L Each
17801347 XL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17861357 Supplier Elma™ Supplier No. 1113840
Only null left
Add to basket
Add to basket
This rinsing sink removes any remaining contamination after cleaning.
  • Important addition to the overall cleaning process
  • Tabletop unit is available in four different sizes – the L and XL models feature an additional side drain
  • Once the cleaning process in the ultrasonic unit is complete, the basket can be inserted directly into the Elma Sink
  • Ensures that any residues and cleaning chemicals are rinsed off thoroughly
  • In conjunction with a suitable rinsing medium, this guarantees optimal cleaning results
Specifications
Type Sink
For Use With (Equipment) Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY
Model M