Learn More
Elma™ Rinsing Sinks
Description
- Important addition to the overall cleaning process
- Tabletop unit is available in four different sizes – the L and XL models feature an additional side drain
- Once the cleaning process in the ultrasonic unit is complete, the basket can be inserted directly into the Elma Sink
- Ensures that any residues and cleaning chemicals are rinsed off thoroughly
- In conjunction with a suitable rinsing medium, this guarantees optimal cleaning results
Specifications
Specifications
| Type | Sink |
| For Use With (Equipment) | Fisherbrand Select, Fisherbrand EASY, Elmasonic P, Elmasonic Select, Elmasonic EASY |
| Model | S |