Learn More
Ansell™ Ringers 085 R-Flex Impact Gloves
Product Code. 17358172 Shop All Ansell Products
Click to view available options
Size:
10
11
12
3X-Large
7
8
9
Unit Size:
1 pair
Product Code. 17358172
Brand: Ansell™ 08511
Only null left
Add to basket
Add to basket
These Ringers 085 R-Flex Impact Gloves provide innovative impact, cut, and cold protection combined with superior dexterity for ultimate comfort in a variety of end uses.
  • The Ringers 085 R-Flex Impact glove with Nitrile Half Dip and Fleece Lining combines protection against impact hazards and the highest levels of cut resistance, with added warmth for application in cold environments.
  • A breathable knit shell developed with Ringers' F3 Technology™ and patented TPR design for superior dexterity and comfort.
  • The sandy finish palm ensures secure grip on wet and dry surfaces, and touchscreen compatible index, middle and thumb tips allow technology interaction.
TRUSTED_SUSTAINABILITY
Specifications
Applications Industrial
Coating Material Nitrile
Color Multicolored
Cut Resistance Rating ANSI A6
Hand Specific
Latex Free Yes
Material HPPE, Nitrile
Product Type Mechanical Protection Gloves
Size 11