Learn More
Brady™ Rigid DuraSleeve™ Vinyl Wire Marking Inserts for M710
Product Code. 17858024
Click to view available options
Dimensions (LxW):
4.4 x 10 mm
4.4 x 12 mm
4.4 x 16 mm
4.4 x 23 mm
4.4 x 30 mm
Color:
White
Yellow
Unit Size:
Pack of 500
Product Code. 17858024
Brand: Brady™ 312002
Only null left
Add to basket
Add to basket
Rigid DuraSleeve™ Wire Marking Inserts for use with Brady Printer M710.
  • Brady™ DuraSleeve™ printable inserts create a durable legend
  • Rigid construction of the DuraSleeve™ wire marking system provides inserts that can easily be inserted into a clean carrier
  • Legend can be changed before or after termination
Specifications
Adhesive Type Non-adhesive
Color White
Finish Gloss
Format Tape
Material Vinyl
Product Type Wire and Cable Markers
Sterility Non-sterile
For Use With (Application) Data and Telecommunications Labeling, Wire and Cable Labeling
For Use With (Equipment) M710, BMP71
Labels per Row 1 Label
Quantity 500 Labels
Temperature Range (English) -40°F to +158°F
Temperature Range (Metric) -40°C to +70°C
Thickness (English) 0.0098 in.
Thickness (Metric) 0.249 mm
Dimensions (LxW) 4.4 x 23 mm
Length (English) 0.17 in.
Width (English) 0.91 in.
Width (Metric) 23 mm
Alternate Part Number M7-23x4.4-7696-WT