Learn More
Epredia™ Richard-Allan Scientific™ Tissue-Loc™ Biopsy Cassettes, White
Product Code. 12617796 Shop All Epredia Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Peach
Pink
Tan
White
Yellow
Quantity:
250/box; 2 boxes
500/Cs.
Unit Size:
Pack of 500
Product Code. 12617796
Brand: Epredia™ 58929
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ensure positive identification from grossing to filing with the three-sided writing surface of Epredia™ Richard-Allan Scientific™ Tissue-Loc™ Biopsy Cassettes.
  • Hinge design ensures specimen safety during processing and easy removal of cover while embedding
  • Base is compatible with metal lids and fits all popular size base molds (metal and disposable plastic)
  • Cassettes are conveniently available assembled or unassembled to facilitate usage in a labeling system or with metal lids
  • Solvent- and decalcifier-resistant and compatible with microwave techniques
  • Different colors allow for easy specimen coding
TRUSTED_SUSTAINABILITY
Specifications
Pore Style Small Square Pore
Quantity 250/box; 2 boxes
Color Tan
Product Type Biopsy Cassette